Iupac Name
N-(2-adamantyl)urea
Inchi Code
1S/C11H18N2O/c12-11(14)13-10-8-2-6-1-7(4-8)5-9(10)3-6/h6-10H,1-5H2,(H3,12,13,14)
InChI key
LQXUPNMWDYTSKT-UHFFFAOYSA-N
Country of Origin
GB
Product Link
https://www.manchesterorganics.com/A17195
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.