ENA769505638
Enamine
ethyl 2,2-diethoxy-3-oxopropanoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
ethyl 2,2-diethoxy-3-oxopropanoate
Inchi Code
1S/C9H16O5/c1-4-12-8(11)9(7-10,13-5-2)14-6-3/h7H,4-6H2,1-3H3
InChI key
MYVHTPMUCKHXOP-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-364564
MSDS
https://www.enaminestore.com/catalog/EN300-364564
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service