ENA497374861
Enamine
2-bromo-1-(1-chlorocyclopropyl)ethan-1-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-bromo-1-(1-chlorocyclopropyl)ethan-1-one
Inchi Code
1S/C5H6BrClO/c6-3-4(8)5(7)1-2-5/h1-3H2
InChI key
PPFFZSUXZGBQDY-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-253224
MSDS
https://www.enaminestore.com/catalog/EN300-253224
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.