Skip to Content
Merck
All Photos(1)

Key Documents

ENA372572521

Enamine

N-[(piperidin-4-yl)methyl]pyridine-2-carboxamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
292.21
MDL number:

Iupac Name

N-(4-piperidinylmethyl)-2-pyridinecarboxamide dihydrochloride

Inchi Code

1S/C12H17N3O.2ClH/c16-12(11-3-1-2-6-14-11)15-9-10-4-7-13-8-5-10;;/h1-3,6,10,13H,4-5,7-9H2,(H,15,16);2*1H

InChI key

LKELOHFWAFAIKB-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-111515

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-111515


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library