ENA321763248
Enamine
[2-(prop-2-en-1-yloxy)phenyl]methanamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
[2-(allyloxy)phenyl]methanamine
Inchi Code
1S/C10H13NO/c1-2-7-12-10-6-4-3-5-9(10)8-11/h2-6H,1,7-8,11H2
InChI key
UUXQXOZZWXRSTG-UHFFFAOYSA-N
storage temp.
RT
Purity
93
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-74303
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-74303
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.