Skip to Content
Merck
All Photos(1)

Documents

C12H316CE76F

Chemcia Scientific, LLC

(3-Amino-4-methoxy-phenyl)-acetonitrile hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
198.65
MDL number:

Iupac Name

2-(3-amino-4-methoxyphenyl)acetonitrile hydrochloride

Inchi Code

1S/C9H10N2O.ClH/c1-12-9-3-2-7(4-5-10)6-8(9)11;/h2-3,6H,4,11H2,1H3;1H

InChI key

HIJRRRFSASERRD-UHFFFAOYSA-N

storage temp.

0-5

Salt Data

HCl

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Light Yellow Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=2268

MSDS

http://www.chemcia.com/product_fl02.asp?id=2268


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service