CH4102152068
ChemBridge Corporation
methyl 2-amino-5-ethylthiophene-3-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 2-amino-5-ethyl-3-thiophenecarboxylate
Inchi Code
1S/C8H11NO2S/c1-3-5-4-6(7(9)12-5)8(10)11-2/h4H,3,9H2,1-2H3
InChI key
YFGOHKLTZXZUPH-UHFFFAOYSA-N
storage temp.
RT
Purity
95%
Country of Origin
KZ
Shipping Temperature
RT
Physical Form
solid
Product Link
https://www.hit2lead.com/building-blocks/4142352
MSDS
https://www.hit2lead.com/sds/4142352_sds.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service