Skip to Content
Merck
All Photos(1)

Documents

V-063

Supelco

Pantothenic acid-13C3,15N hemicalcium salt solution

100 μg/mL (90:10 Methanol:Water, (as free acid)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C6H1615NO5 · 0.5Ca
CAS Number:
Molecular Weight:
242.24
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (90:10 Methanol:Water, (as free acid))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

An internal standard suitable for quantitation of pantothenic acid levels and testing applications including clinical and diagnostic testing or quality control testing of vitamin supplements, fortified foods or pharmaceutical preparations. Pantothenic acid, also known as Vitamin B5, is an essential water-soluble B vitamin that is involved in the coenzyme A (CoA) synthesis.

Application


  • Enzyme inhibition studies: Stable isotope-labeled pantothenic acid is used in X-ray crystallography and NMR studies to understand the mechanism of homotropic inhibition in enzyme reactions, specifically pantothenate synthetase. Such studies are essential for drug design targeting metabolic pathways, enhancing the effectiveness of treatments involving vitamin B5 pathways (Chakrabarti et al., 2010).

  • NMR structural analysis: The labeled compound aids in the NMR structural elucidation of enzymes such as ketopantoate reductase, helping to determine its role in biological systems. This analysis is crucial for developing drugs that can manipulate these enzymatic pathways, impacting treatments for conditions requiring metabolic modulation (Headey et al., 2008).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service