Skip to Content
Merck
All Photos(1)

Key Documents

T-904

Supelco

Trimipramine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2
CAS Number:
Molecular Weight:
294.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CN(C)CC(C)CN1C2=C(C=CC=C2)CCC3=C1C=CC=C3

InChI

1S/C20H26N2/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22/h4-11,16H,12-15H2,1-3H3

InChI key

ZSCDBOWYZJWBIY-UHFFFAOYSA-N

General description

Trimipramine is a tricyclic antidepressant prescribed for treatment of depression, insomnia, and chronic pain or as an adjunctive therapy for alcohol or opioid withdrawal. This certified solution standard is applicable for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC-MS/MS or GC/MS. Trimipramine is marketed under the trade names Surmontil®, Rhotrimine, and Stangyl.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Surmontil is a registered trademark of Pliva, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britta Haenisch et al.
Psychopharmacology, 217(2), 289-295 (2011-04-13)
The antidepressant trimipramine shows an atypical pharmacological profile and its mechanism of action is still obscure. The present study investigated whether trimipramine and three of its metabolites interact with targets of other antidepressants, namely, the human monoamine transporters for noradrenaline
Donald F Smith et al.
Psychopharmacology, 195(1), 131-138 (2007-07-27)
Molecular tools are needed for assessing anti-depressant actions by positron emission tomography (PET) in the living human brain. This study determined whether [(11)C]mirtazapine is an appropriate molecular tool for use with PET to estimate the magnitude of neuroreceptor occupancy produced
M T Jafari et al.
Analytical and bioanalytical chemistry, 399(10), 3555-3564 (2011-02-08)
A novel method based on three-phase hollow fiber microextraction technique (HF-LPME) coupled with electrospray ionization-ion mobility spectrometry (ESI-IMS) was developed for the simultaneous determination of two antidepressant drugs (trimipramine and desipramine) in urine and plasma samples. The effects of various
Thomas Gelbrich et al.
Acta crystallographica. Section C, Crystal structure communications, 68(Pt 2), o65-o70 (2012-02-07)
Polymorph (Ia) of eldoral [5-ethyl-5-(piperidin-1-yl)barbituric acid or 5-ethyl-5-(piperidin-1-yl)-1,3-diazinane-2,4,6-trione], C(11)H(17)N(3)O(3), displays a hydrogen-bonded layer structure parallel to (100). The piperidine N atom and the barbiturate carbonyl group in the 2-position are utilized in N-H···N and N-H···O=C hydrogen bonds, respectively. The structure
Ali R Fakhari et al.
Electrophoresis, 33(3), 506-515 (2012-01-31)
A sensitive, simple and reproducible method was developed for preconcentration and determination of trimipramine (TPM) enantiomers in biological samples using electromembrane extraction combined with cyclodextrin-modified capillary electrophoresis (CE). During the extraction, TPM enantiomers migrated from a 5 mL sample solution through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service