Skip to Content
Merck
All Photos(1)

Key Documents

T-167

Supelco

Temazepam-D5 glucuronide lithium salt solution

100 μg/mL in methanol ((as free carboxylate)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€428.00

€428.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€428.00

About This Item

Empirical Formula (Hill Notation):
C22H15D5ClN2O8Li
Molecular Weight:
487.83
UNSPSC Code:
41116107
NACRES:
NA.24

€428.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol ((as free carboxylate))

format

single component solution

storage temp.

−20°C

General description

This stable-labeled internal standard is suitable for quantification of temazepam glucuronide by LC/MS or GC/MS for clinical toxicology, forensic analysis, hydrolysis controls, or urine drug testing applications. The major urinary and blood metabolite of temazepam, an intermediate-acting 3-hydroxy benzodiazepine prescribed for the short-term treatment of sleeplessness in patients with difficulties remaining asleep.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service