Skip to Content
Merck
All Photos(1)

Key Documents

SCD-003

Supelco

Dibenz[a,h]anthracene

vial of 1 g, analytical standard, Cerilliant®

Synonym(s):

1,2:5,6-Dibenzanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912416
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available. Contact Technical Service

grade

analytical standard

packaging

vial of 1 g

manufacturer/tradename

Cerilliant®

bp

524 °C (lit.)

mp

262-265 °C (lit.)

application(s)

environmental
forensics and toxicology

format

neat

storage temp.

room temp

SMILES string

c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23

InChI

1S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14H

InChI key

LHRCREOYAASXPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenz[a,h]anthracene belongs to the class of polynuclear aromatic hydrocarbons (PAHs). PAHs are a group of well-known environmental pollutants with both mutagenic and carcinogenic properties. They are released into the environment by the pyrolysis of organic materials.[1]

Application

Dibenz[a,h]anthracene may be used as an analytical reference standard for the determination of the analyte in cane sugar,[1] marine samples,[2] bulk precipitation and runoff waters[3] by chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of polycyclic aromatic hydrocarbons in bulk precipitation and runoff waters in an urban region (Poland)
Grynkiewicz M, et al.
Atmospheric Environment, 36(2), 361-369 (2002)
Determination of polycyclic aromatic hydrocarbons in cane sugar
Tfouni SAV and Toledo MCF
Food Control, 18(8), 948-952 (2007)
Determination of polynuclear aromatic hydrocarbons in marine samples of Siokolo Fishing Settlement
Anyakora C, et al.
Journal of Chromatography A, 1073(1-2), 323-330 (2005)
M Shou et al.
Archives of biochemistry and biophysics, 328(1), 201-207 (1996-04-01)
The metabolic activation of the potent carcinogen dibenzo[a,h]anthracene (DB[a,h]A) was investigated with recombinant human cytochrome P450 enzymes 1A2, 2B6, 2C8, 2C9, 2E1, 3A3, 3A4, and 3A5 expressed in hepatoma G2 cells and with 14 different human liver microsomes. Three dihydrodiols
B Z Zhong et al.
Mutation research, 326(2), 147-153 (1995-02-01)
Benz[a]anthracene (BA), dibenz[a,h]anthracene (DBA) and dibenzo[a,i]pyrene (DBP) are polycyclic aromatic hydrocarbons (PAHs) found in incomplete combustion products of fossil fuels, coal tar, and other organic materials. Workers in related industries may be exposed to these chemicals by inhalation. The information

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service