Skip to Content
Merck
All Photos(1)

Key Documents

S-039

Supelco

JWH-018 4-Hydroxypentyl metabolite-D5 (indole-D5) solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

JWH-018 4-Hydroxypentyl metabolite-d5 (indole-d5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H18D5NO2
Molecular Weight:
362.48
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[n]1(c2c(c(c1[2H])C(=O)c3c4c(ccc3)cccc4)c(c(c(c2[2H])[2H])[2H])[2H])CCCC(O)C

InChI

1S/C24H23NO2/c1-17(26)8-7-15-25-16-22(20-12-4-5-14-23(20)25)24(27)21-13-6-10-18-9-2-3-11-19(18)21/h2-6,9-14,16-17,26H,7-8,15H2,1H3/i4D,5D,12D,14D,16D

InChI key

LLFBQQHYZJTRAS-NXZSVEOISA-N

General description

A stable-labeled internal standard of JWH-018 4-Hydroxypentyl metabolite, also known as the Omega minus one-OH metabolite of JWH-018. The 4-Hydroxypentyl analog is the major urinary metabolite of JWH-018, a synthetic cannabinoid found in the herbal mixture Spice.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service