Skip to Content
Merck
All Photos(1)

Key Documents

R-023

Supelco

Rufinamide solution

1.0 mg/mL in acetonitrile: water (9:1), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€150.00

€150.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€150.00

About This Item

Empirical Formula (Hill Notation):
C10H8F2N4O
CAS Number:
Molecular Weight:
238.19
UNSPSC Code:
41116107
NACRES:
NA.24

€150.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile: water (9:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC(=O)c1cn(Cc2c(F)cccc2F)nn1

InChI

1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)4-16-5-9(10(13)17)14-15-16/h1-3,5H,4H2,(H2,13,17)

InChI key

POGQSBRIGCQNEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An antiepileptic drug and anticonvulsant, Rufinamide is approved for the treatment of partial seizures associated with Lennox-Gastaut syndrome in adults and children 4 years and older. Rufinamide is marketed as Banzel® in the US and Inovelon® in the EU.

Application

An antiepileptic drug and anticonvulsant, Rufinamide is approved for the treatment of partial seizures associated with Lennox-Gastaut syndrome in adults and children 4 years and older.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service