Skip to Content
Merck
All Photos(1)

Documents

M-154

Supelco

MDPBP hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

3’,4’-Methylenedioxy-alpha-pyrrolidinobutiophenone hydrochloride solution, 3′,4′-Methylenedioxy-alpha-pyrrolidinobutiophenone hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO3 · HCl
CAS Number:
Molecular Weight:
297.78
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCC(C(C1=CC=C2C(OCO2)=C1)=O)N3CCCC3.Cl

InChI

1S/C15H19NO3.ClH/c1-2-12(16-7-3-4-8-16)15(17)11-5-6-13-14(9-11)19-10-18-13;/h5-6,9,12H,2-4,7-8,10H2,1H3;1H

InChI key

ATCKCJHEOJFKJW-UHFFFAOYSA-N

General description

A novel designer drug first developed in the 1960′s as a research chemical, MDPBP first emerged as a legal high in Europe in 2009. This synthetic cathinone and bath salt is often sold in the US and other countries under the name "NRG-1" as a mixture with other substituted cathinone derivatives such as flephedrone, pentylone, and MDPV.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service