Skip to Content
Merck
All Photos(1)

Key Documents

M-145

Supelco

Milnacipran hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€194.00

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€194.00

About This Item

Empirical Formula (Hill Notation):
C15H23ClN2O
CAS Number:
Molecular Weight:
282.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CCN(CC)C([C@]1(C2=CC=CC=C2)C[C@@H]1CN)=O.Cl

InChI

1S/C15H22N2O.ClH/c1-3-17(4-2)14(18)15(10-13(15)11-16)12-8-6-5-7-9-12;/h5-9,13H,3-4,10-11,16H2,1-2H3;1H/t13-,15+;/m1./s1

InChI key

XNCDYJFPRPDERF-PBCQUBLHSA-N

General description

Milnacipran, sold under the brand names Savella®, Ixel®, Dalcipran®, or Toledomin, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the treatment of depression. In 2009, the US Food and Drug Administration (FDA) approved milnacipran for management of the chronic pain disorder fibromyalgia, making it only the third drug approved in the US for this indication besides duloxetine and pregabalin.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dalcipran is a registered trademark of Pierre Fabre Medicament
Ixel is a registered trademark of Pierre Fabre Medicament
Savella is a registered trademark of Forest Laboratories, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service