Skip to Content
Merck
All Photos(1)

Key Documents

M-016

Supelco

(±)-Methamphetamine-D8 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D8H7N
CAS Number:
Molecular Weight:
157.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Product M-016 is not currently sold in your country. Contact Technical Service

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])NC(C([2H])([2H])[2H])([2H])C([H])([2H])C1=C([H])C([H])=C([H])C([H])=C1[H]

InChI

1S/C10H15N/c1-9(11-2)8-10-6-4-3-5-7-10/h3-7,9,11H,8H2,1-2H3/i1D3,2D3,8D,9D

InChI key

MYWUZJCMWCOHBA-URFKORASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes known by the street names "meth," "ice," and "glass". This internal standard is suitable for quantitation of methamphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, sports testing, forensic analysis, or isotope dilution methods. The dextrorotatory isomer of methamphetamine is a pharmaceutical drug sold under the trade name Desoxyn® for treatment of ADHD and obesity.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Desoxyn is a registered trademark of Abbott Laboratories Corp.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service