Skip to Content
Merck
All Photos(1)

Key Documents

I-025

Supelco

Interference Mix 3 (For Qualitative Use Only)

50-1000 μg/mL each component in acetonitrile, analytical standard, Cerilliant®

Synonym(s):

Qualitative Interference Mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product I-025 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

form

liquid

composition

Aripiprazole, 100 μg/mL
Lacosamide, 100 μg/mL
Oxcarbazepine, 100 μg/mL
(+)-Propoxyphene, 50 μg/mL
Rufinamide, 100 μg/mL
Warfarin, 1000 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

50-1000 μg/mL each component in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

A multi-component solution standard of routinely monitored over-the-counter (OTC) and prescription drugs (+)-propoxyphene; aripiprazole; lacosamide; oxcarbazepine; rufinamide; and warfarin. This standard is for qualitative use only in the evaluation of interferences between the mix components and target analytes of interest in a testing laboratory′s analytical methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1A - STOT RE 2

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service