Skip to Content
Merck
All Photos(1)

Key Documents

H-050

Supelco

Hydromorphone-D6 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13NO3D6
CAS Number:
Molecular Weight:
291.37
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product H-050 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1N([C@@H]2Cc3ccc(O)c4O[C@H]5C(=O)CC[C@@H]2[C@]5(c34)C1([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C17H19NO3/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18/h2,4,10-11,16,19H,3,5-8H2,1H3/t10-,11+,16-,17-/m0/s1/i1D3,6D2,7D/t7?,10-,11+,16-,17-

InChI key

WVLOADHCBXTIJK-LJWMZWAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydromorphone, marketed under brand names including Palladone and Dilaudid®, is a very potent opiate used as an analgesic for treating pain or as a cough suppressant for painful coughing resulting from continued bronchial irritation. This Certified Spiking Solution® is suitable for isotope dilution methods or quantitation of hydromorphone levels by GC/MS or LC/MS for clinical toxicology, pain prescription monitoring, urine drug testing, or pharmaceutical research applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Dilaudid is a registered trademark of Purdue Pharma L.P.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service