Skip to Content
Merck
All Photos(1)

Key Documents

G-017

Supelco

Gabapentin-D10 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€1,690.00

€1,690.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€1,690.00

About This Item

Empirical Formula (Hill Notation):
C9D10H7NO2
CAS Number:
Molecular Weight:
181.30
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€1,690.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])C(CN)(CC(O)=O)C([2H])([2H])C1([2H])[2H]

InChI

1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)/i1D2,2D2,3D2,4D2,5D2

InChI key

UGJMXCAKCUNAIE-YXALHFAPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sold under the trade names Neurontin®, Nupentin, or Gabarone, gabapentin is an anticonvulsant drug used primarily for treatment of seizures, fibromyalgia, and chronic neuropathic pain. This internal standard is suitable for GC/MS or LC/MS testing or isotope dilution methods of gabapentin in forensic analysis, clinical toxicology, pain prescription monitoring, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Neurontin is a registered trademark of Warner-Lambert Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service