Skip to Content
Merck
All Photos(1)

Key Documents

F-057

Supelco

Fenproporex-D5 hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Labeled Fenproporex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11D5N2 · HCl
Molecular Weight:
229.76
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product F-057 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C(CC(C)NCCC#N)C([2H])=C([2H])C([2H])=C1[2H].Cl

InChI

1S/C12H16N2.ClH/c1-11(14-9-5-8-13)10-12-6-3-2-4-7-12;/h2-4,6-7,11,14H,5,9-10H2,1H3;1H/i2D,3D,4D,6D,7D;

InChI key

ZBRZSJUFJUMKIM-MKCOZUEUSA-N

General description

This stable isotope labeled Snap-N-Shoot® Reference Solution is suitable for quantification of fenproporex in a variety of LC/MS or GC/MS applications from forensic analysis and urine drug testing and to clinical toxicology and sports testing. Fenproporex, also known as Perphoxene and used as an appetite suppressant for the treatment of obesity, is a stimulant drug of the phenethylamine and amphetamine classes that produces amphetamine as a metabolite. Fenproporex is designated a Schedule IV controlled substance in the US pursuant to the Controlled Substances Act and is also listed as banned substance by the World Anti-Doping Agency.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service