Skip to Content
Merck
All Photos(1)

Documents

ERR-005S

Supelco

RDX solution

10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant®

Synonym(s):

Hexahydro-1,3,5-trinitro-1,3,5-triazine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N6O6
CAS Number:
Molecular Weight:
222.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 5 mL

manufacturer/tradename

Cerilliant®

concentration

10 mg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

O=[N+]([O-])N1CN([N+]([O-])=O)CN([N+]([O-])=O)C1

InChI

1S/C3H6N6O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H2

InChI key

XTFIVUDBNACUBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT RE 2 Inhalation

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Ping Chen et al.
Applied and environmental microbiology, 78(21), 7798-7800 (2012-08-28)
Whole-genome sequencing, transcriptomic analyses, and metabolic reconstruction were used to investigate Gordonia sp. strain KTR9's ability to catabolize a range of compounds, including explosives and steroids. Aspects of this mycolic acid-containing actinobacterium's catabolic potential were experimentally verified and compared with
Anat Bernstein et al.
Environmental science & technology, 47(1), 479-484 (2012-12-12)
The explosive Hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) is known to be degraded aerobically by various isolates of the Rhodococcus species, with denitration being the key step, mediated by Cytochrome P450. Our study aimed at gaining insight into the RDX degradation mechanism by Rhodococcus
Karl K Irikura
The journal of physical chemistry. A, 117(10), 2233-2241 (2013-02-05)
The explosive nitramine RDX (1,3,5-trinitrohexahydro-s-triazine) is thought to decompose largely by homolytic N-N bond cleavage, among other possible initiation reactions. Density-functional theory (DFT) calculations indicate that the resulting secondary aminyl (R2N·) radical can abstract an oxygen atom from NO2 or
Thomas J Bruno et al.
Journal of chromatography. A, 1286, 192-199 (2013-03-13)
In previous work, dynamic headspace vapor collection on short, porous layer open tubular (PLOT) capillary columns maintained at low temperature was introduced. In this paper, that metrology is extended with the introduction of a small in situ pyrolysis platform that
Muhammad Imran Khan et al.
Journal of microbiology and biotechnology, 22(10), 1311-1323 (2012-10-19)
In the present work, current knowledge on the potential fate, microbial degradation, and toxicity of hexahydro- 1,3,5-trinitro-1,3,5-triazine (RDX) was thoroughly reviewed, focusing on the toxicological assessment of a variety of potential RDX degradation pathways in bacteria and fungi. The present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service