Skip to Content
Merck
All Photos(1)

Key Documents

E-015S

Supelco

Ethyl-beta-D-glucuronide solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Ethyl-β-D-glucuronide

Sign Into View Organizational & Contract Pricing

Select a Size

E-015S-1ML
€245.00

€245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
E-015S-1ML
€245.00

About This Item

Empirical Formula (Hill Notation):
C8H14O7
CAS Number:
Molecular Weight:
222.19
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€245.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O[C@H]1[C@H](O)[C@@H](C(O)=O)O[C@@H](OCC)[C@@H]1O

InChI

1S/C8H14O7/c1-2-14-8-5(11)3(9)4(10)6(15-8)7(12)13/h3-6,8-11H,2H2,1H3,(H,12,13)/t3-,4-,5+,6-,8+/m0/s1

InChI key

IWJBVMJWSPZNJH-UQGZVRACSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl-ß-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alcohol consumption. This Snap-N-Spike® Reference Solution is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service