Skip to Content
Merck
All Photos(1)

Key Documents

D-108

Supelco

Dihydromorphine-3ß-D-glucuronide solution

1.0 mg/mL in methanol: water (1:1), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29NO9
CAS Number:
Molecular Weight:
463.48
UNSPSC Code:
41116107

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol: water (1:1)

format

single component solution

storage temp.

−20°C

SMILES string

CN1[C@H](C2)C3[C@@]4(CC1)C5=C2C=CC(O[C@H]6[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O6)=C5O[C@H]4[C@@H](O)CC3

InChI

1S/C23H29NO9/c1-24-7-6-23-10-3-4-12(25)20(23)32-18-13(5-2-9(14(18)23)8-11(10)24)31-22-17(28)15(26)16(27)19(33-22)21(29)30/h2,5,10-12,15-17,19-20,22,25-28H,3-4,6-8H2,1H3,(H,29,30)/t10?,11-,12+,15+,16+,17-,19+,20+,22-,23+/m1/s1

InChI key

KCPRJVDBLLJBMF-RURUVCMVSA-N

Application

A Certified Snap-N-Shoot® solution standard for LC/MS or GC/MS applications with dihydromorphine-3β-D-glucuronide, such as pharmaceutical research, forensic analysis, urine drug testing or clinical toxicology. Dihydromorphine-3β-D-glucuronide is a major urinary of dihydromorphine, an opiate commonly used for the management of moderate to severe pain. 

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service