Skip to Content
Merck
All Photos(1)

Key Documents

C-249

Supelco

Cariprazine-D8 solution

100 μg/mL (Acetonitrile: Water with 1% 1M HCl (50:50) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€156.00

€156.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€156.00

About This Item

Empirical Formula (Hill Notation):
C21H24D8Cl2N4O
CAS Number:
Molecular Weight:
435.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€156.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Acetonitrile: Water with 1% 1M HCl (50:50) (v/v))

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service