Skip to Content
Merck
All Photos(1)

Key Documents

C-089

Supelco

1-(3-Chlorophenyl)piperazine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

1-(3-Chlorophenyl)piperazine ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14Cl2N2
CAS Number:
Molecular Weight:
233.14
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product C-089 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[Cl-].Clc1cc(ccc1)[N+H]2CCNCC2

InChI

1S/C10H13ClN2.ClH/c11-9-2-1-3-10(8-9)13-6-4-12-5-7-13;/h1-3,8,12H,4-7H2;1H

InChI key

MHXPYWFZULXYHT-UHFFFAOYSA-N

General description

1-(3-Chlorophenyl)-piperazine, also known as meta-chlorophenylpiperazine or mCPP, is a designer drug analog of aryl-substituted piperazines. A stimulant, serotonin antagonist and reuptake inhibitor (SARU), mCPP can also be detected as a primary urinary metabolite of the antidepressant drug trazadone. This certified solution standard is suitable for use in piperazine testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Cao et al.
Journal of analytical toxicology, 39(5), 335-346 (2015-04-03)
A novel LC-MS-MS assay that simultaneously detects and quantitates 78 drugs and metabolites was developed and validated for chronic pain management. Urine specimen was diluted and mixed with internal standards (ISs) before injected into LC-MS-MS. Seventy-two analytes were detected with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service