Skip to Content
Merck
All Photos(1)

Key Documents

A-113

Supelco

AH-7921 hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22Cl2N2O · HCl
CAS Number:
Molecular Weight:
365.73
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product A-113 is not currently sold in your country. Contact Technical Service

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)
USDEA Schedule I

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=C(Cl)C=CC(C(NCC2(N(C)C)CCCCC2)=O)=C1.Cl

InChI

1S/C16H22Cl2N2O.ClH/c1-20(2)16(8-4-3-5-9-16)11-19-15(21)12-6-7-13(17)14(18)10-12;/h6-7,10H,3-5,8-9,11H2,1-2H3,(H,19,21);1H

InChI key

GYMKEMDHKUDSBK-UHFFFAOYSA-N

General description

This certified Snap-N-Spike® Solution is suitable for use as starting material in calibrators and controls for AH-7921 LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications. AH-7921 is an opioid designer drug with 80% the potency of morphine when administered orally. First discovered in 2013, AH-7921 is an active ingredient of synthetic cannabis found in Japan.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Hayes et al.
British journal of pharmacology, 79(3), 731-736 (1983-07-01)
The effects of mu and kappa-opiate receptor agonists were studied in a variety of tests in the mouse designed to correspond to clinical side-effects in man. These included sedation, decrease in pupil diameter, Straub tail, decrease in body temperature, decrease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service