Skip to Content
Merck
All Photos(1)

Key Documents

LM1003

Avanti

21:0-22:6 PC

Avanti Research - A Croda Brand LM1003, methanol solution

Synonym(s):

1-heneicosanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H90NO8P
CAS Number:
Molecular Weight:
876.24
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1003-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1003

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CCCCCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C51H90NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-51(54)60-49(48-59-61(55,56)58-46-45-52(3,4)5)47-57-50(53)43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h8,10,14,16,20,22,26,28,32,34,38,40,49H,6-7,9,11-13,15,17-19,21,23

InChI key

MBNMYHRROGWUAS-AWYUTHGESA-N

General description

Phosphatidylcholine (PC) is an abundant phospholipid in the pulmonary surfactant.

Application

21:0-22:6 PC or 1-heneicosanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphocholine has been used as a lipid standard/LIPID MAPS (LM) internal standard for phospholipid quantification/analysis using mass spectrometry (MS) and liquid chromatography (LC)-MS.

Packaging

2 mL Amber Glass Sealed Ampule (LM1003-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Per Larsson et al.
Respiratory physiology & neurobiology, 243, 39-46 (2017-05-16)
Exhaled particles constitute a micro-sample of respiratory tract lining fluid. Inhalations from low lung volumes generate particles in small airways by the airway re-opening mechanism. Forced exhalations are assumed to generate particles in central airways by mechanisms associated with high
Penelope Dimas et al.
eLife, 8 (2019-05-08)
Oligodendrocytes (OLs) support neurons and signal transmission in the central nervous system (CNS) by enwrapping axons with myelin, a lipid-rich membrane structure. We addressed the significance of fatty acid (FA) synthesis in OLs by depleting FA synthase (FASN) from OL
Laura Montani et al.
The Journal of cell biology, 217(4), 1353-1368 (2018-02-13)
Myelination calls for a remarkable surge in cell metabolism to facilitate lipid and membrane production. Endogenous fatty acid (FA) synthesis represents a potentially critical process in myelinating glia. Using genetically modified mice, we show that Schwann cell (SC) intrinsic activity
Ulrike Bruning et al.
Cell metabolism, 28(6), 866-880 (2018-08-28)
The role of fatty acid synthesis in endothelial cells (ECs) remains incompletely characterized. We report that fatty acid synthase knockdown (FASNKD) in ECs impedes vessel sprouting by reducing proliferation. Endothelial loss of FASN impaired angiogenesis in vivo, while FASN blockade reduced
Alexander Fauland et al.
Journal of lipid research, 52(12), 2314-2322 (2011-10-01)
This work aims to combine chromatographic retention, high mass resolution and accuracy, MS/MS spectra, and a package for automated identification and quantitation of lipid species in one platform for lipidomic analysis. The instrumental setup elaborated comprises reversed-phase HPLC coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service