Skip to Content
Merck
All Photos(1)

Key Documents

870731P

Avanti

18:1 (n12) Coenzyme A

Avanti Research - A Croda Brand 870731P, powder

Synonym(s):

(6Z-octadecenoyl) Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€366.00

€366.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€366.00

About This Item

Empirical Formula (Hill Notation):
C39H77N10O17P3S
CAS Number:
Molecular Weight:
1083.07
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€366.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870731P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870731P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCC/C=C\CCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C39H68N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-30(48)67-23-22-41-29(47)20-21-42-37(51)34(50)39(2,3)25-60-66(57,58)63-65(55,56)59-24-28-33(62-64(52,53)54)32(49)38(61-28)46-27-45-31-35(40)43-26-44-36(31)46;;;/h14-15,26-28,32-34,38,49-50

InChI key

MWMWRTNFCHQAGB-GFCZCHAHSA-N

General description

18:1 (n12) Coenzyme A, also known as (6Z-octadecenoyl) coenzyme A, is a coenzyme A derivative of (6Z)-Octadec-6-enoic acid.

Packaging

5 mL Amber Glass Screw Cap Vial (870731P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service