Skip to Content
Merck
All Photos(1)

Key Documents

870600P

Avanti

PAzePC

Avanti Research - A Croda Brand 870600P, powder

Synonym(s):

1-palmitoyl-2-azelaoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€126.00

€126.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€126.00

About This Item

Empirical Formula (Hill Notation):
C33H64NO10P
CAS Number:
Molecular Weight:
665.84
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€126.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (870600P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870600P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCC(O)=O)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C33H64NO10P/c1-5-6-7-8-9-10-11-12-13-14-15-18-21-24-32(37)41-28-30(29-43-45(39,40)42-27-26-34(2,3)4)44-33(38)25-22-19-16-17-20-23-31(35)36/h30H,5-29H2,1-4H3,(H-,35,36,39,40)/t30-/m1/s1

InChI key

GHQQYDSARXURNG-SSEXGKCCSA-N

General description

1-palmitoyl-2-azelaoyl-sn-glycero-3-phosphocholine (PAzePC) structure contains an anionic carboxyl group at the end of the short, oxidized acyl chain.[1]

Application

1-palmitoyl-2-azelaoyl-sn-glycero-3-phosphocholine (PazePC) may be used as an oxidized lipid dopant in liposomes for examining the link between the permeation of H2O2 and the structure of lipid membranes.[2] It may also be used as lysosomal phospholipase A2 substrate in the study of structural basis for Zn2+ mediated inhibition of lysosomal phospholipase A2.[3]

Biochem/physiol Actions

1-palmitoyl-2-azelaoyl-sn-glycero-3-phosphocholine (PazePC) is a stable lipid oxidation product.[1] It serves as an inter-H2O2 permeation promoter.[2] It is the most common drug target for antipsychotic and antimicrobial agents in cells under oxidative stress at inflammatory sites.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870600P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renee A Bouley et al.
Biochemistry, 58(13), 1709-1717 (2019-03-05)
Lysosomal phospholipase A2 (LPLA2/PLA2G15) is a key enzyme involved in lipid homeostasis and is characterized by both phospholipase A2 and transacylase activity and by an acidic pH optimum. Divalent cations such as Ca2+ and Mg2+ have previously been shown to
Himanshu Khandelia et al.
Biophysical journal, 96(7), 2734-2743 (2009-04-08)
In oxidative environments, biomembranes contain oxidized lipids with short, polar acyl chains. Two stable lipid oxidation products are PoxnoPC and PazePC. PoxnoPC has a carbonyl group, and PazePC has an anionic carboxyl group pendant at the end of the short
Yuya Ouchi et al.
Scientific reports, 9(1), 12497-12497 (2019-08-31)
H2O2 permeation through a cell membrane significantly affects living organisms, and permeation is controlled by the physico-chemical nature of lipids and other membrane components. We investigated the molecular relationship between H2O2 permeation and lipid membrane structure using three oxidized lipids.
Mooud Amirkavei et al.
Biochimica et biophysica acta, 1858(2), 264-273 (2015-12-15)
In order to obtain molecular level insight into the biophysics of the apoptosis promoting phospholipid 1-palmitoyl-2-azelaoyl-sn-glycero-3-phosphocholine (PazePC) we studied its partitioning into different lipid phases by isothermal titration calorimetry (ITC). To aid the interpretation of these data for PazePC, we
Cintia Kawai et al.
The journal of physical chemistry. B, 118(41), 11863-11872 (2014-09-24)
Fluorescence quenching of lipid-bound pyrene was used to assess the binding of cytochrome c (cyt c) to liposomes that mimic the inner mitochondrial membrane (IMM) POPC/DOPE/TOCL, with the conditions that it did or did not contain oxidized phosphatidylcholine molecules, i.e.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service