Skip to Content
Merck
All Photos(1)

Documents

870140P

Avanti

16:0 Dodecanylamine PE

Avanti Research - A Croda Brand 870140P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H97N2O9P
CAS Number:
Molecular Weight:
889.28
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870140P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870140P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCCCCCCCC[NH3+])=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C49H97N2O9P/c1-3-5-7-9-11-13-15-17-19-23-27-31-35-39-48(53)57-44-46(60-49(54)40-36-32-28-24-20-18-16-14-12-10-8-6-4-2)45-59-61(55,56)58-43-42-51-47(52)38-34-30-26-22-21-25-29-33-37-41-50/h46H,3-45,50H2,1-2H3,(H,51,52)(H,55,56)/t46-/m1/s1

InChI key

MFAZBSZBKVFZAN-YACUFSJGSA-N

General description

16:0 Dodecanylamine PE or 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine), belongs to the family of head group modified functionalized lipids. Functionalized lipids mainly act as reporter molecules.

Application

16:0 Dodecanylamine PE may be used as a co-immobilized lipid molecule to increase the stability and sensitivity of a superoxide anion radical (O2•–) microbiosensor.

Packaging

5 mL Amber Glass Screw Cap Vial (870140P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Aminur Rahman et al.
Analytical chemistry, 84(15), 6654-6660 (2012-07-19)
Enhanced stability and sensitivity of a superoxide anion radical (O(2)(•-)) microbiosensor were achieved through the sequential immobilization of lipid and cytochrome c (Cyt c) covalently bonded onto a conducting polymer layer that showed a clear quasi-reversible direct electron transfer (DET)
Functionalized lipids and surfactants for specific applications
Kepczynski M and Rog T
Biochimica et Biophysica Acta - Biomembranes, 1858(10), 2362-2379 (2016)
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service