Skip to Content
Merck
All Photos(1)

Key Documents

860903P

Avanti

17:0-17:1-17:0 D5 TG

Avanti Research - A Croda Brand 860903P, powder

Synonym(s):

1,3(d5)-diheptadecanoyl-2-(10Z-heptadecenoyl)-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€163.00

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C54H97D5O6
CAS Number:
Molecular Weight:
852.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860903P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860903P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OC(CCCCCCCCCCCCCCCC)=O)(OC(CCCCCCCC/C=C\CCCCCC)=O)C([2H])([2H])OC(CCCCCCCCCCCCCCCC)=O

InChI

1S/C54H102O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-52(55)58-49-51(60-54(57)48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)50-59-53(56)47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h21,24,51H,4-20,22-23,25-50H2,1-3H3/b24-21-/i49D2,50D2,51D

InChI key

OWYYELCHNALRQZ-ADIIQMQPSA-N

Application

17:0-17:1-17:0 D5 TG is suitable for use as a lipid standard to identify lipid metabolites by ultra-performance liquid chromatography - tandem mass spectrometer (UPLC-MS).[1][2] It is also suitable for use as an internal standard to measure triglyceride concentration in horse adipose tissue, human plasma and liver tissue by QTRAP mass spectrometer.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860903P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive qualification and quantification of triacylglycerols with specific fatty acid chain composition in horse adipose tissue, human plasma and liver tissue
Guan M, et al.
Talanta, 172, 206-214 (2017)
The Effects of Age and Reproduction on the Lipidome of Caenorhabditis elegans
Wan QL, et al.
Oxidative Medicine and Cellular Longevity, 2019 (2019)
Microbial ecology on solar panels in Berkeley, CA, United States
Porcar M, et al.
Frontiers in Microbiology, 9 (2018)
Bingpeng Yan et al.
International journal of molecular sciences, 20(23) (2019-11-30)
Enterovirus A71 (EV-A71) and coxsackievirus A16 (CV-A16) are the most common causes of hand, foot, and mouth disease. Severe EV-A71 and CV-A16 infections may be associated with life-threatening complications. However, the pathogenic mechanisms underlying these severe clinical and pathological features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service