Skip to Content
Merck
All Photos(1)

Key Documents

860854P

Avanti

C24:0 Galactosyl(β)ceramide(d18:1/24:0)

Avanti Research - A Croda Brand (860854P), powder

Synonym(s):

D-galactosyl-β-1,1’-N-lignoceroyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€392.00

€392.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€392.00

About This Item

Empirical Formula (Hill Notation):
C48H93NO8
CAS Number:
Molecular Weight:
812.25
UNSPSC Code:
12352100
NACRES:
NA.25

€392.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (860854P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand (860854P)

lipid type

deuterated lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C48H93NO8/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-44(52)49-41(40-56-48-47(55)46(54)45(53)43(39-50)57-48)42(51)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h35,37,41-43,45-48,50-51,53-55H,3-34,36,38-40H2,1-2H3,(H,49,52)/b37-35+/t41-

InChI key

POQRWMRXUOPCLD-HIIAJUEOSA-N

General description

β-Galactosyl ceramide (β-GalCer) is a simple glycosphingolipid. It is made up of lipophilic sphingosine & hydrophilic galactose attached to sphingosine via ether linkage. Β-GalCer is present in the myelin sheath of the central and peripheral nervous system. It has five analogs including 6a-6e.[1]

Biochem/physiol Actions

β-Galactosyl ceramide (β-GalCer) regulates the activity of β-glucocerebrosidase. It also plays a role as cell surface receptors for bacterial and viral toxins.[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis, and evaluation of beta-galactosylceramide mimics promoting beta-glucocerebrosidase activity in keratinocytes
Fukunaga K, et al.
Bioorganic & Medicinal Chemistry Letters, 13(5), 813-815 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service