Skip to Content
Merck
All Photos(1)

Documents

860842P

Avanti

18:0(2R-OH) Sulfo GalCer

Avanti Research - A Croda Brand 860842P, powder

Synonym(s):

3-O-sulfo-D-galactosyl-β1-1′-N-[2"(R)-hydroxystearyl]-D-erythro-sphingosine (ammonium salt), (synthetic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84N2O12S
CAS Number:
Molecular Weight:
841.19
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860842P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860842P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC([C@H](O)CCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](OS([O-])(=O)=O)[C@H]([C@H]1CO)O.[NH4+]

General description

18:0(2R-OH) Sulfo GalCer, also known as 3-O-sulfo-D-galactosyl-β1-1′-N-[2"(R)-hydroxystearyl]-D-erythro-sphingosine, is a sulfatide that belongs to the class of sphingolipids. It is highly found in membranous myelin sheath formed around nerve axons in gangliosides. This sulfated galactosylceramide contains 3-O-sulfo-β-d-galactose moiety linked to transmembrane-embedded unique ceramide containing 18C long chain base fatty acid (stearic acid) with 2′-hydroxyl group in R configuration.

Biochem/physiol Actions

Sulfatides are involved in various cellular processes such as protein trafficking, signal transduction and neuronal cell differentiation. Accumulation of sulfoglycolipids (mainly sulfo-galactosylceramide, S-GalCer) in lysosomes, due to the inherited deficiency of arylsulfatase A (ASA), causes metachromatic leukodystrophy in humans. In mice, deficiency of sulfatide causes paralysis due to demyelination.

Packaging

5 mL Amber Glass Screw Cap Vial (860842P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vongsavanh Phongsisay et al.
Molecular immunology, 63(2), 595-599 (2014-08-31)
Axonal Guillain-Barré syndrome (GBS) is an autoimmune neuropathy characterized by limb weakness and/or paralysis due to the presence of autoantibodies against brain glycolipids. The immune receptors that recognize these autoimmune targets have not been described. In this study, 12 C-type
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 116(2), 223-249 (2008)
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 116(2), 223-249 (2008)
R Lüllmann-Rauch et al.
Histochemistry and cell biology, 116(2), 161-169 (2001-10-31)
The inherited deficiency of arylsulfatase A (ASA) causes lysosomal accumulation of sulfoglycolipids (mainly sulfo-galactosylceramide, S-GalCer ) and leads to metachromatic leukodystrophy in humans. Among visceral organs, kidneys are particularly affected. In the present study, the regional distribution and temporal development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service