Skip to Content
Merck
All Photos(1)

Key Documents

860828P

Avanti

18:1(2S-OH) Ceramide

Avanti Research - A Croda Brand 860828P, powder

Synonym(s):

N-(2′-(S)-hydroxyoleoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€299.00

€299.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€299.00

About This Item

Empirical Formula (Hill Notation):
C36H69NO4
CAS Number:
Molecular Weight:
579.94
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€299.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860828P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860828P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@@H](O)CCCCCC/C=C\CCCCCCCC)=O)CO

InChI key

OGMXRWJJGUINGB-GRQDMSQNSA-N

General description

18:1(2S-OH) Ceramide is a 2S hydroxylated oleic acid-containing sphingolipid.[1] hFA- ceramides are mainly found in epidermis.[2]

Biochem/physiol Actions

Ceramides containing hydroxy fatty acids play a vital role in epidermal permeability barrier function.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860828P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Guo et al.
Journal of lipid research, 53(7), 1327-1335 (2012-04-21)
FA 2-hydroxylase (FA2H) is an NAD(P)H-dependent enzyme that initiates FA α oxidation and is also responsible for the biosynthesis of 2-hydroxy FA (2-OH FA)-containing sphingolipids in mammalian cells. The 2-OH FA is chiral due to the asymmetric carbon bearing the
Hiroko Hama
Biochimica et biophysica acta, 1801(4), 405-414 (2009-12-23)
2-Hydroxy fatty acids (hFA) are important components of a subset of mammalian sphingolipids. The presence of hFA in sphingolipids is best described in the nervous system, epidermis, and kidney. However, the literature also indicates that various hFA-sphingolipids are present in
Ana B Herrero et al.
Cancer research, 68(23), 9779-9787 (2008-12-03)
PM02734 is a novel synthetic antitumor drug that is currently in phase I clinical trials. To gain some insight into its mode of action, we used the yeast Saccharomyces cerevisiae as a model system. Treatment of S. cerevisiae with PM02734
Nathan L Alderson et al.
Journal of lipid research, 50(6), 1203-1208 (2009-01-28)
Sphingolipids are ubiquitous components of eukaryotic cells that regulate various cellular functions. In many cell types, a fraction of sphingolipids contain 2-hydroxy fatty acids, produced by fatty acid 2-hydroxylase (FA2H), as the N-acyl chain of ceramide [hydroxyl fatty acid (hFA)-sphingolipids].
Zdzislaw M Szulc et al.
Bioorganic & medicinal chemistry, 18(21), 7565-7579 (2010-09-21)
A straightforward method for the simultaneous preparation of (2S,3R,2'R)- and (2S,3R,2'S)-2'-hydroxy-ceramides (2'-OHCer) from (2S,3R)-sphingosine acetonide precursors and racemic mixtures of 2-hydroxy fatty acids (2-OHFAs) is described. The obtained 2'-OH-C4-, -C6-, -C12-, -C16-Cer and 2'-OH-C6-dhCer pairs of diastereoisomers were characterized thoroughly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service