Skip to Content
Merck
All Photos(1)

Key Documents

860672P

Avanti

C22 Ceramide (d14:1/22:0)

Avanti Research - A Croda Brand, powder

Synonym(s):

N-behenoyl-D-erythro-sphingosine (C14 base)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,250.00

€1,250.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€1,250.00

About This Item

Empirical Formula (Hill Notation):
C36H71NO3
CAS Number:
Molecular Weight:
565.95
UNSPSC Code:
12352100
NACRES:
NA.25

€1,250.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

1 ea of 10 mg (860672P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids
deuterated lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCC

InChI

1S/C36H71NO3/c1-3-5-7-9-11-13-14-15-16-17-18-19-20-21-22-24-26-28-30-32-36(40)37-34(33-38)35(39)31-29-27-25-23-12-10-8-6-4-2/h29,31,34-35,38-39H,3-28,30,32-33H2,1-2H3,(H,37,40)/b31-29+/t34-,35+/m0/s1

InChI key

ZEQGGWNOJHBYRD-NXCSZAMKSA-N

Application

C22 Ceramide (d14:1/22:0) has been used as a standard for measuring sphingomyelins and ceramides through a multiple reaction monitoring (MRM) method by ultrafast-liquid chromatography (UFLC)-mass spectrometry (MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of ER stress on sphingolipid levels and apoptotic pathways in retinal pigment epithelial cells
Afcsar E, et al.
Redox Biology, 101430-101430 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service