Skip to Content
Merck
All Photos(1)

Key Documents

860618P

Avanti

CER6-2′R(d9)

Avanti Research - A Croda Brand 860618P, powder

Synonym(s):

N-(2′-(R)-hydroxypalmitoyl(d9)) D-ribo-phytosphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€382.00

€382.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€382.00

About This Item

Empirical Formula (Hill Notation):
C34H60D9NO5
CAS Number:
Molecular Weight:
580.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€382.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860618P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860618P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@]([H])(CCCCCCCCCCCCCC)[C@](O)([H])[C@@]([H])(NC([C@H](O)CCCCCCCCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O)CO

InChI

1S/C34H69NO5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-31(37)33(39)30(29-36)35-34(40)32(38)28-26-24-22-20-18-16-14-12-10-8-6-4-2/h30-33,36-39H,3-29H2,1-2H3,(H,35,40)/t30-,31+,32+,33-/m0/s1/i2D3,4D2,6D2,8D2

InChI key

RHIXBFQKTNYVCX-OCIMHASQSA-N

Application

CER6-2′R(d9) or N-(2′-(R)-hydroxypalmitoyl(d9)) D-ribo-phytosphingosine has been used as an internal standard for quantification of stratum corneum (SC) ceramides in humans and mice by liquid chromatography-mass spectrometry (LC-MS)/MS.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860618P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Momoko Kawana et al.
Journal of lipid research, 61(6), 884-895 (2020-04-09)
Ceramides are the predominant lipids in the stratum corneum (SC) and are crucial components for normal skin barrier function. Although the composition of various ceramide classes in the human SC has been reported, that in mice is still unknown, despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service