Skip to Content
Merck
All Photos(1)

Key Documents

860590P

Avanti

C18:1 Lactosyl(β) Ceramide (d18:1/18:1)

Avanti Research - A Croda Brand 860590P, powder

Synonym(s):

D-lactosyl-β-1,1′ N-oleyol-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€223.00

€223.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€223.00

About This Item

Empirical Formula (Hill Notation):
C48H89NO13
CAS Number:
Molecular Weight:
888.22
UNSPSC Code:
12352211
NACRES:
NA.25

€223.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860590P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860590P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI key

YVBUQOZKCCPFCZ-HSVHIEHGSA-N

Application

C18:1 Lactosyl(β) Ceramide (d18:1/18:1) or D-lactosyl-β-1,1′ N-oleyol-D-erythro-sphingosine has been used as a standard for structural analysis of glycosphingolipid containing unsaturated fatty acyl chain using ozone-induced dissociation-mass spectrometry (OzID-MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860590P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodell C Barrientos et al.
Journal of the American Society for Mass Spectrometry, 28(11), 2330-2343 (2017-08-24)
Glycosphingolipids are essential biomolecules widely distributed across biological kingdoms yet remain relatively underexplored owing to both compositional and structural complexity. While the glycan head group has been the subject of most studies, there is paucity of reports on the lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service