Skip to Content
Merck
All Photos(1)

Key Documents

860579P

Avanti

Galactosyl(β) Dimethyl Sphingosine (d18:1)

Avanti Research - A Croda Brand 860579P, powder

Synonym(s):

N,N-dimethyl-D-galactosyl-β1-1′-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€429.00

€429.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€429.00

About This Item

Empirical Formula (Hill Notation):
C26H51NO7
CAS Number:
Molecular Weight:
489.69
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€429.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860579P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860579P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@](N(C)C)([H])CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C26H51NO7/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(29)20(27(2)3)19-33-26-25(32)24(31)23(30)22(18-28)34-26/h16-17,20-26,28-32H,4-15,18-19H2,1-3H3/b17-16+/t20-,21+,22+,23-,24-,25+,26+/m0/s1

InChI key

SJWBHOQBARABKH-QHRJNYSLSA-N

General description

Galactosyl(β) Dimethyl Sphingosine (d18:1) is a galactosylated derivative of dimethyl sphingosine, which is a methylated sphingosine. It belongs to the category of glycosylated sphingolipids. Glycosylated sphingolipids are basic building blocks of rafts (cell membrane compartments).[1]

Biochem/physiol Actions

Glycosylated sphingolipids are implicated in the stimulation of receptor mediated signaling events.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860579P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Baumruker et al.
Seminars in immunology, 14(1), 57-63 (2002-03-09)
Over the last decade evidence has accumulated that sphingolipids are important and specific signalling molecules for cell-to-cell communication (mediator function) as well as for intracellular signalling processes (second messenger function). In addition, glycosylated sphingolipids are essential building blocks of rafts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service