Skip to Content
Merck
All Photos(1)

Documents

860577P

Avanti

C24 Lactosyl(β) Ceramide (d18:1/24:0)

Avanti Research - A Croda Brand 860577P, powder

Synonym(s):

D-lactosyl-β-1,1′ N-lignoceroyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H103NO13
CAS Number:
Molecular Weight:
974.39
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860577P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860577P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

Application

C24 Lactosyl(β) Ceramide (d18:1/24:0) or D-lactosyl-β-1,1′ N-lignoceroyl-D-erythro-sphingosine has been used as a standard to determine the fragmentation pattern of glycosphingolipid (GSLs) using liquid chromatography coupled with mass spectrometry (LC/MS) in three strains of Skeletonema.

Packaging

5 mL Amber Glass Screw Cap Vial (860577P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Zhao et al.
Rapid communications in mass spectrometry : RCM, 27(13), 1535-1547 (2013-06-01)
Glycosphingolipids (GSLs) are important components of the cell membrane; however, little is known about GSLs in microalgae. We analyzed GSLs in three strains of Skeletonema microalgae by liquid chromatography coupled with mass spectrometry. Total lipid was extracted from Skeletonema microalgae.
Annemieke de Jong et al.
Nature immunology, 15(2), 177-185 (2013-12-24)
T cells autoreactive to the antigen-presenting molecule CD1a are common in human blood and skin, but the search for natural autoantigens has been confounded by background T cell responses to CD1 proteins and self lipids. After capturing CD1a-lipid complexes, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service