Skip to Content
Merck
All Photos(1)

Key Documents

860556C

Avanti

18:1-12:0 Biotin DG

Avanti Research - A Croda Brand 860556C

Synonym(s):

1-oleoyl-2-[12-biotinyl(aminododecanoyl)]-sn-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€626.00

€626.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€626.00

About This Item

Empirical Formula (Hill Notation):
C43H77N3O7S
CAS Number:
Molecular Weight:
780.15
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€626.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (860556C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860556C

concentration

1 mg/mL (860556C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](CO)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C43H77N3O7S/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-23-30-40(49)52-34-36(33-47)53-41(50)31-24-21-18-15-13-16-19-22-27-32-44-39(48)29-26-25-28-38-42-37(35-54-38)45-43(51)46-42/h9-10,36-38,42,47H,2-8,11-35H2,1H3,(H,44,48)(H2,45,46,51)/b10-9-/t36-,37-,38-,42-/m0/s1

InChI key

DPGSXRFXXINSBQ-QYFPMEDBSA-N

General description

18:1-12:0 Biotin DG or 1-oleoyl-2-[12-biotinyl(aminododecanoyl)]-sn-glycerol is a triglyceride derivative wherein aminododecyl forms an amide bond with biotin to yield a biotinylated triglyceride.

Packaging

5 mL Amber Glass Screw Cap Vial (860556C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leslie A McCauliff et al.
eLife, 8 (2019-10-04)
Unesterified cholesterol accumulation in the late endosomal/lysosomal (LE/LY) compartment is the cellular hallmark of Niemann-Pick C (NPC) disease, caused by defects in the genes encoding NPC1 or NPC2. We previously reported the dramatic stimulation of NPC2 cholesterol transport rates to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service