Skip to Content
Merck
All Photos(1)

Key Documents

860552P

Avanti

Biotin S1P

Avanti Research - A Croda Brand 860552P, powder

Synonym(s):

omega-biotinyl D-erythro-sphingosine-1-phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€435.00

€435.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€435.00

About This Item

Empirical Formula (Hill Notation):
C28H53N4O7PS
CAS Number:
Molecular Weight:
620.78
UNSPSC Code:
12352211
NACRES:
NA.25

€435.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (860552P-100ug))

manufacturer/tradename

Avanti Research - A Croda Brand 860552P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]1(CCCCC(NCCCCCCCCCCCCC/C=C/[C@@]([H])(O)[C@]([NH3+])([H])COP([O-])(O)=O)=O)SC[C@@]2([H])[C@]1([H])NC(N2)=O

InChI

1S/C28H53N4O7PS/c29-22(20-39-40(36,37)38)24(33)16-12-10-8-6-4-2-1-3-5-7-9-11-15-19-30-26(34)18-14-13-17-25-27-23(21-41-25)31-28(35)32-27/h12,16,22-25,27,33H,1-11,13-15,17-21,29H2,(H,30,34)(H2,31,32,35)(H2,36,37,38)/b16-12+/t22-,23-,24+,25-,27-/m0/s1

InChI key

JHROEHVYJZWKKP-RPRIXWAOSA-N

General description

Biotinylated sphingosine 1-phosphate (Biotin S1P) is produced by the phosphorylation of biotinyl sphingosine by recombinant human sphingosine kinase 1 and 2 (hSK1 and hSK2). Biotinyl sphingosine is derived from sphingosine and it contains a biotin unit at the ω-terminal of its fatty-acid chain.[1]

Application

Biotinylated sphingosine 1-phosphate (Biotin S1P) is suitable to be used as an antigen to preabsorb murine monoclonal antibody, LT1002.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (860552P-100ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An assay for sphingosine kinase activity using biotinylated sphingosine and streptavidin-coated membranes
Roberts J, et al.
Analytical biochemistry, 331(1), 122-129 (2004)
Immunohistochemical detection of sphingosine-1-phosphate and sphingosine kinase-1 in human tissue samples
Sphingosine-1-Phosphate, 55-67 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service