Skip to Content
Merck
All Photos(1)

Key Documents

860536P

Avanti

Sphinganine-1-Phosphate (d18:0)

Avanti Research - A Croda Brand 860536P, powder

Synonym(s):

D-erythro-sphinganine-1-phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€240.00

€240.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€240.00

About This Item

Empirical Formula (Hill Notation):
C18H40NO5P
CAS Number:
Molecular Weight:
381.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€240.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860536P-1mg)
pkg of 1 × 5 mg (860536P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860536P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](CCCCCCCCCCCCCCC)(O)[C@]([NH3+])([H])COP([O-])(O)=O

InChI

1S/C18H40NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)17(19)16-24-25(21,22)23/h17-18,20H,2-16,19H2,1H3,(H2,21,22,23)/t17-,18+/m0/s1

InChI key

YHEDRJPUIRMZMP-ZWKOTPCHSA-N

General description

Sphinganine-1-Phosphate (SA1P) is a bioactive lipid present in mammals. It is produced from the sphingoid base, sphinganine in the presence of sphingosine kinase (SPHK).[1]

Application

Sphinganine-1-Phosphate (d18:0) has been used as a reference standard in ultra performance liquid chromatography coupled to a mass spectrometry.[2] It is suitable for use as an internal standard for ultra high performance liquid chromatography for the quantification of blood and muscle sphingolipid.[1] Sphinganine-1-phosphate (d18:0) is also suitable for use as standard in liquid chromatography tandem mass spectrometry.[3]

Biochem/physiol Actions

Sphinganine-1-Phosphate (SA1P) is an important marker for cancer and immune system disorders studies. It elicits protective functionality in hepatic ischemia.[3] Elevated levels of SA1P is observed in hepatocellular carcinoma.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (860536P-1mg)
5 mL Amber Glass Screw Cap Vial (860536P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum sphingolipidomic analyses reveal an upregulation of C16-ceramide and sphingosine-1-phosphate in hepatocellular carcinoma
Grammatikos G, et al.
Testing, 7(14), 18095-18095 (2016)
Accurate quantification of sphingosine-1-phosphate in normal and Fabry disease plasma, cells and tissues by LC-MS/MS with 13C-encoded natural S1P as internal standard
Mirzaian M, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 459(5), 36-44 (2016)
Exercise increases sphingoid base-1-phosphate levels in human blood and skeletal muscle in a time-and intensity-dependent manner
Baranowski M, et al.
European Journal of Applied Physiology, 115(5), 993-1003 (2015)
Metabolomic insights into system-wide coordination of vertebrate metamorphosis
Ichu T, et al.
BMC Developmental Biology, 14(1), 5-5 (2014)
Katja Jakobi et al.
International journal of molecular sciences, 21(7) (2020-04-05)
Hepatocellular carcinoma (HCC) shows a remarkable heterogeneity and is recognized as a chemoresistant tumor with dismal prognosis. In previous studies, we observed significant alterations in the serum sphingolipids of patients with HCC. This study aimed to investigate the in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service