Skip to Content
Merck
All Photos(1)

Documents

860532P

Avanti

C8 Ceramide-1-Phosphate (d18:1/8:0)

N-octanoyl-ceramide-1-phosphate (ammonium salt), powder

Synonym(s):

Ammonium (2S,3R,4E)-3-hydroxy-2-(octanoylamino)octadec-4-en-1-yl hydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H55N2O6P
CAS Number:
Molecular Weight:
522.70
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860532P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860532P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI

1S/C26H52NO6P.H3N/c1-3-5-7-9-10-11-12-13-14-15-16-18-19-21-25(28)24(23-33-34(30,31)32)27-26(29)22-20-17-8-6-4-2;/h19,21,24-25,28H,3-18,20,22-23H2,1-2H3,(H,27,29)(H2,30,31,32);1H3/b21-19+;/t24-,25+;/m0./s1

InChI key

WDZBNEWCUHJJDF-AAMXSQEYSA-N

General description

C8 Ceramide-1-Phosphate (d18:1/8:0) or N-octanoyl-ceramide-1-phosphate is a synthetic short-chain ceramide.
Ceramide-1-phosphate is synthesized from ceramide by the phosphorylation reaction catalyzed by ceramide kinase (CerK).

Application

C8 Ceramide-1-Phosphate (d18:1/8:0) has been used as one of the authentic standards to validate selected metabolites in metabolomic study of vertebrate metamorphosis.
C8 Ceramide-1-Phosphate (d18:1/8:0) or N-octanoyl-ceramide-1-phosphate has been used:
  • in lipid preparation to treat endothelial colony-forming cells (ECFCs) for nuclear morphology analysis
  • for the validation of C8 ceramide-1-phosphate metabolite to study its effects on the metabolic pathways of Rana catesbeiana during metamorphosis
  • to screen lipids that rapidly and reversibly alter transepithelial electrical resistance (TER) or tight junction (TJ) permeability in epithelial tissue

Biochem/physiol Actions

Ceramide-1-phosphate (C1P), as a lipid derived secondary messenger plays a vital role in various cellular processes such as inflammation, cell proliferation, differentiation and apoptosis. It is mitogenic for fibroblasts. C1P is also implicated in the modulation of macrophage chemotaxis. It promotes cell-survival by inhibiting the enzyme acid sphingomyelinase (SMase).

Packaging

5 mL Amber Glass Screw Cap Vial (860532P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide-1-phosphate in cell survival and inflammatory signaling
Gomez-Munoz, A, et al.
Sphingolipids as Signaling and Regulatory Molecules, 118-130 (2010)
Ceramide and ceramide 1-phosphate in health and disease
Arana L, et al.
Lipids in Health and Disease, 9(1), 15-15 (2010)
Hebe Agustina Mena et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(10), e219-e232 (2019-08-23)
Ceramide 1-phosphate (C1P) is a bioactive sphingolipid highly augmented in damaged tissues. Because of its abilities to stimulate migration of murine bone marrow-derived progenitor cells, it has been suggested that C1P might be involved in tissue regeneration. In the present
Metabolomic insights into system-wide coordination of vertebrate metamorphosis
Ichu T, et al.
BMC Developmental Biology, 14(1), 5-5 (2014)
Antonio Gómez-Muñoz et al.
Journal of lipid research, 45(1), 99-105 (2003-10-03)
It was reported previously that ceramide-1-phosphate (Cer-1-P) is mitogenic for fibroblasts (Gómez-Muñoz, A., P. A. Duffy, A. Martin, L. O'Brien, H-S. Byun, R. Bittman, and D. N. Brindley. 1995. Mol. Pharmacol. 47: 883-889; Gómez-Muñoz, A., L. M. Frago, L. Alvarez

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service