Skip to Content
Merck
All Photos(2)

Key Documents

860462P

Avanti

N-C16-deoxysphinganine

N-palmitoyl-1-deoxysphinganine (m18:0/16:0), powder

Synonym(s):

N-hexadecanoyl-1-deoxysphinganine (m18:0/16:0); N-C16-1-deoxyDHCer; 110960

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€308.00

€308.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€308.00

About This Item

Empirical Formula (Hill Notation):
C34H69NO2
CAS Number:
Molecular Weight:
523.92
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€308.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860462P-1mg)
pkg of 1 × 5 mg (860462P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860462P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

General description

Commonly referred to as 1-deoxydihydroceramide (1-deoxyDHCer), this product is the N-acylated form of 1-deoxysphinganine, a potent inhibitor of sphingolipid metabolism. The biological activity of 1-deoxyDHCer is not clearly understood at this time.
N-C16-deoxysphinganine Commonly referred to as 1-deoxydihydroceramide (1-deoxyDHCer), is the N-acylated form of 1-deoxysphinganine,[1] a potent inhibitor of sphingolipid metabolism. The N-acyl group can be 16, 20 and 24 carbon chain.[2] N-acylsphinganines (dihydroceramides) are synthesized by the acylation of sphingoid bases in the presence of ceramide synthases (CerS).[3]

Biochem/physiol Actions

N-acylsphinganines (dihydroceramides) levels are lower during ceramide synthase inhibition. They are intermediates of ceramide and dihydrosphingolipids synthesis. Treatment of MCF7 cancer cells with fenretinide alters sphingolipid metabolism.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860462P-1mg)
5 mL Amber Glass Screw Cap Vial (860462P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingolipidomics: a valuable tool for understanding the roles of sphingolipids in biology and disease
Merrill AH, et al.
Journal of Lipid Research, 50, S97-S102 (2009)
Ceramide synthase inhibition by fumonisin B1 causes accumulation of 1-deoxysphinganine a novel category of bioactive 1-deoxysphingoid bases and 1-deoxydihydroceramides biosynthesized by mammalian cell lines and animals
Zitomer NC, et al.
The Journal of Biological Chemistry, 284(8), 4786-4795 (2009)
Thematic Review Series: Sphingolipids. Biodiversity of sphingoid bases (?sphingosines?) and related amino alcohols
Pruett ST, et al.
Journal of Lipid Research, 49(8), 1621-1639 (2008)
Noemi Jiménez-Rojo et al.
Biophysical journal, 107(12), 2850-2859 (2014-12-18)
Ceramides and dihydroceramides are N-acyl derivatives of sphingosine and sphinganine, respectively, which are the major sphingoid-base backbones of mammals. Recent studies have found that mammals, like certain other organisms, also produce 1-deoxy-(dihydro)ceramides (1-deoxyDHCers) that contain sphingoid bases lacking the 1-hydroxyl-
Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service