Skip to Content
Merck
All Photos(2)

Key Documents

860399P

Avanti

16:0-d31-18:1 PC

1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

110918

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€159.00
25 MG
€301.00
100 MG
€772.00

€159.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€159.00
25 MG
€301.00
100 MG
€772.00

About This Item

Empirical Formula (Hill Notation):
C42H51NO8PD31
CAS Number:
Molecular Weight:
791.27
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

€159.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860399P-10mg)
pkg of 1 × 100 mg (860399P-100mg)
pkg of 1 × 25 mg (860399P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860399P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1/i2D3,7D2,9D2,11D2,13D2,15D2,17D2,19D2,22D2,24D2,26D2,28D2,30D2,32D2,34D2

InChI key

WTJKGGKOPKCXLL-KHCKXXADSA-N

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31-18:1 PC may be used in the preparation of mitochondrial mimetic liposomes.[1] It may also be used as an internal standard for the quantification of lipids using electrospray ionization tandem mass spectrometry (ESI-MS/MS).[2]

Packaging

5 mL Clear Glass Sealed Ampule (860399P-100mg)
5 mL Clear Glass Sealed Ampule (860399P-10mg)
5 mL Clear Glass Sealed Ampule (860399P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delineating the rules for structural adaptation of membrane-associated proteins to evolutionary changes in membrane lipidome
Makarova M, et al.
Current Biology (2020)
Rita Touti et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 151, 18-31 (2020-04-11)
Multi-lamellar liposomes (MLLs), prepared by shearing a lamellar phase composed of lipids (phosphatidylcholine) and surfactant (Tween 80®), were designed to control their size, charge and elasticity, the key parameters known to influence liposomes penetration through skin. Their size was tuned
Surface-binding to cardiolipin nanodomains triggers cytochrome c pro-apoptotic peroxidase activity via localized dynamics
Li M, et al.
Structure, 27(5), 806-815 (2019)
Kotaro Hama et al.
Journal of lipid research, 61(4), 523-536 (2020-02-23)
X-linked adrenoleukodystrophy (X-ALD) is an inherited disorder caused by deleterious mutations in the ABCD1 gene. The ABCD1 protein transports very long-chain FAs (VLCFAs) from the cytosol into the peroxisome where the VLCFAs are degraded through β-oxidation. ABCD1 dysfunction leads to
Alessandra Luchini et al.
Analytical chemistry, 92(1), 1081-1088 (2019-11-27)
In vitro characterization of membrane proteins requires experimental approaches providing mimics of the microenvironment that proteins encounter in native membranes. In this context, supported lipid bilayers provide a suitable platform to investigate membrane proteins by a broad range of surface-sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service