Skip to Content
Merck
All Photos(2)

Key Documents

860336P

Avanti

MAPCHO-12-d38

dodecyl phosphocholine-d38, powder

Synonym(s):

DPC-D38

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€470.00

€470.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€470.00

About This Item

Empirical Formula (Hill Notation):
C17D38NO4P
CAS Number:
Molecular Weight:
389.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€470.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (860336P-100mg)
pkg of 1 × 200 mg (860336P-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860336P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OC([2H])(C([N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])[2H])[2H])(OC([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H]

InChI

1S/C17H38NO4P/c1-5-6-7-8-9-10-11-12-13-14-16-21-23(19,20)22-17-15-18(2,3)4/h5-17H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2

InChI key

QBHFVMDLPTZDOI-DHIOKLJKSA-N

Application

MAPCHO-12-d38 has been used in the preparation of drug complexes used as a sample during nuclear magnetic resonance (NMR) spectroscopy.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860336P-100mg)
5 mL Amber Glass Screw Cap Vial (860336P-200mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
MAPCHO is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Small Molecule Targeting the Transmembrane Domain of Death Receptor p75NTR Induces Melanoma Cell Death and Reduces Tumor Growth
Goh E, et al.
Cell Chemical Biology, 25(12), 1485-1494 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service