Skip to Content
Merck
All Photos(2)

Key Documents

860202C

Avanti

16:0 Phosphatidylbutanol

1,2-dipalmitoyl-sn-glycero-3-phosphobutanol (sodium salt), chloroform

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphobutanol (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€138.00

€138.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€138.00

About This Item

Empirical Formula (Hill Notation):
C39H76O8PNa
CAS Number:
Molecular Weight:
726.98
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€138.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (860202C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860202C

concentration

10 mg/mL (860202C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCC)OCCCC

InChI

1S/C39H77O8P.Na/c1-4-7-10-12-14-16-18-20-22-24-26-28-30-32-38(40)44-35-37(36-46-48(42,43)45-34-9-6-3)47-39(41)33-31-29-27-25-23-21-19-17-15-13-11-8-5-2;/h37H,4-36H2,1-3H3,(H,42,43);/q;+1/p-1/t37-;/m1./s1

InChI key

CZFXRJUSZGHONR-GKEJWYBXSA-M

General description

Phosphatidylbutanol is a low polarity phospholipid, which can be produced by phospholipase D (PLD) specific transphosphatidylation reaction of phosphatidylcholine (PC) and butanol.[1]

Application

16:0 Phosphatidylbutanol or 1,2-dipalmitoyl-sn-glycero-3-phosphobutanol (sodium salt) has been used to analyze the in vivo activity of phospholipase D (PLD) in Taxus cuspidate using liquid chromatography electrospray ionization tandem mass spectrometric (LC/ESI-MS/MS).[2][3][4]

Biochem/physiol Actions

Phosphatidylbutanol is used as a biomarker in phospholipase D activity assays.[1]

Packaging

5 mL Clear Glass Sealed Ampule (860202C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-pei Han et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(2), 623-630 (2008-10-17)
Lipid signaling involved in mechanotransduction processes in response to shear stress in plants remained elusive. To understand the responses of phospholipids in shear stress-induced mechanotransduction, a lipidomic approach was employed to profile phospholipid species of Taxus cuspidata cells under laminar
Song Yang et al.
Biochimica et biophysica acta, 1771(5), 600-612 (2007-04-13)
A comparative lipidomics approach was employed to investigate the changes in membrane phospholipids during the procession of cellular development and apoptosis of two plant cell lines, Taxus cuspidata and Taxus chinensis var. mairei. Analysis of lipids by LC/ESI/MS(n) showed more
Song Yang et al.
Chemistry and physics of lipids, 159(1), 13-20 (2009-05-12)
Degradation of membrane phospholipids is associated with apoptotic responses, but the signaling development of this degradation is not well understood. Cerium (Ce(4+)), an important rare earth element, induces cellular apoptosis and taxol biosynthesis in Taxus cuspidata suspension cultures. Here, using
Mikhail Vyssotski et al.
Lipids, 44(4), 381-389 (2008-12-17)
High-performance TLC and (31)P-NMR were assessed as methods of observing the presence of numerous low polarity phospholipids: bis-phosphatidic acid (BPA), semi-lyso bis-phosphatidic acid (SLBPA), N-acyl phosphatidylethanolamine (NAPE), N-(1,1-dimethyl-3-oxo-butyl)-phosphatidylethanolamine (diacetone adduct of PE, DOBPE), N-acetyl PE, phosphatidylmethanol (PM), phosphatidylethanol (PEt), phosphatidyl-n-propanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service