860129W
Avanti
C18:0-d7 Fuc-GM1 Ceramide (d18:1/18:0-d7)
from synthetic
Synonym(s):
C18:0-d7 Fuc-GM1 Ganglioside ((Fuca1,2)Galb1,3GalNAcb1,4(Neu5Aca2,3)Galb1,4Glcb-Ceramide(d18:1/18:0-d7))
About This Item
Recommended Products
biological source
synthetic
Assay
≥99% (TLC)
form
methanol solution
packaging
pkg of 1 × 100 μg (860129W-100UG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
sphingolipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])CCCCCCCCCCCCCCC(=O)N[C@@H](CO[C@@H]1OC(CO)[C@@H](O[C@@H]2OC(CO)[C@H](O[C@@H]3OC(CO)[C@H](O)C(O[C@@H]4OC(CO)[C@H](O)C(O)C4O[C@@H]4OC(C)[C@@H](O)C(O)C4O)C3NC(C)=O)C(O[C@@]3(CC(O)[C@@H](NC(C)=O)C(O3)[C@H](O)[C@H](O)CO)C(O)=O)C2O)C(O)C1O)C(O)\C=C\CCCCCCCCCCCCC
Application
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service