Skip to Content
Merck
All Photos(1)

Documents

857521P

Avanti

N-Octanoyl-L-homoserine lactone

(S)-N-(2-Oxotetrahydrofuran-3-yl)octanamide, powder

Synonym(s):

N-[(3S)-Tetrahydro-2-oxo-3-furanyl]octanamide, C8-HSL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO3
CAS Number:
Molecular Weight:
227.30
Beilstein:
8148282
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (857521P-10MG)
pkg of 1 × 50 mg (857521P-50MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bacterial lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C1OCC[C@@H]1NC(CCCCCCC)=O

InChI

1S/C12H21NO3/c1-2-3-4-5-6-7-11(14)13-10-8-9-16-12(10)15/h10H,2-9H2,1H3,(H,13,14)/t10-/m0/s1

InChI key

JKEJEOJPJVRHMQ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Octanoyl-L-homoserine lactone (C8-HSL) is a N-acyl homoserine lactone (N-acyl-HSL). It is highly nonpolar.

Biochem/physiol Actions

N-Octanoyl-L-homoserine lactone (C8-HSL) is produced by many gram-negative bacteria like Pseudomonas fluorescens, Ralstonia solanacearum, Vibrio fischeri, and Yersinia pseudotuberculosis. In Escherichia coli, C8-HSL is encoded by the gene Ytb1. Generally, these homoserine lactones (HSLs) are quorum sensing signaling molecules. HSLs play a key role in biofilm formation. They function as an autoinducer molecule and involved in cell to cell communication depending on cell density population.
Quorum-sensing signal generation

Packaging

5 mL Amber Glass Screw Cap Vial (857521P-10MG and 857521P-50MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of N-acylhomoserine lactones produced by bacteria isolated from industrial cooling water systems.
Okutsu N, et al.
Sensors, 16(1), 44-44 (2016)
A hierarchical quorum-sensing system in Yersinia pseudotuberculosis is involved in the regulation of motility and clumping.
Atkinson S, et al.
Molecular Microbiology, 33(6), 1267-1277 (1999)
P D Shaw et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(12), 6036-6041 (1997-06-10)
Many Gram-negative bacteria regulate gene expression in response to their population size by sensing the level of acyl-homoserine lactone signal molecules which they produce and liberate to the environment. We have developed an assay for these signals that couples separation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service