Skip to Content
Merck
All Photos(2)

Key Documents

857392O

Avanti

(R)-FTY720-OMe

(R)-2-amino-2-(methoxymethyl)-4-(4-octylphenyl)butan-1-ol, neat oil

Synonym(s):

(R)-FTY720-methyl ether; ROME

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€166.00

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€166.00

About This Item

Empirical Formula (Hill Notation):
C20H35NO2
CAS Number:
Molecular Weight:
321.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mg (857392O-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857392O

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

General description

(R)-FTY720-OMe is the analogue of FTY720.[1]
(R)-FTY720-OMe is a structural analogue of FTY720 which acts as a specific competitive inhibitor of sphingosine kinase 2 (SK2). Furthermore, (R)-FTY720-OMe does not inhibit sphingosine kinase 1 (SK1) activity.

Biochem/physiol Actions

(R)-FTY720-OMe helps to block DNA synthesis and actin rearrangement induced by sphingosine 1-phosphate (S1P) in MCF-7 breast cancer cells. It can also reduce sphingosine kinase 2 (SK2) expression and prevent DNA synthesis in HEK 293 cells. (R)-FTY720-OMe has anti-breast cancer potential.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857392O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Tonelli et al.
British journal of pharmacology, 168(6), 1497-1505 (2012-11-02)
Sphingosine kinase catalyses the formation of sphingosine 1-phosphate and is linked with androgen receptor signalling in prostate cancer cells. Therefore, we investigated the effect of sphingosine kinase inhibitors on androgen receptor expression. Androgen-sensitive LNCaP cells were treated with SKi (2-(p-hydroxyanilino)-4-(p-chlorophenyl)thiazole)
(R)-FTY720 methyl ether is a specific sphingosine kinase 2 inhibitor: effect on sphingosine kinase 2 expression in HEK 293 cells and actin rearrangement and survival of MCF-7 breast cancer cells
Lim KG, et al.
Cellular Signalling, 23(10), 1590-1595 (2011)
Keng Gat Lim et al.
Cellular signalling, 23(10), 1590-1595 (2011-05-31)
Sphingosine kinase 2 (SK2) catalyses the conversion of sphingosine to the bioactive lipid sphingosine 1-phosphate (S1P). We report here, the stereospecific synthesis of an analogue of FTY720 called (R)-FTY720-OMe, which we show is a competitive inhibitor of SK2. (R)-FTY720-OMe failed
David G Watson et al.
Cellular signalling, 25(4), 1011-1017 (2013-01-15)
Two isoforms of sphingosine kinase, SK1 and SK2, catalyze the formation of the bioactive lipid sphingosine 1-phosphate (S1P) in mammalian cells. We have previously shown that treatment of androgen-sensitive LNCaP prostate cancer cells with a non-selective SK isoform inhibitor, 2-(p-hydroxyanilino)-4-(p-chlorophenyl)thiazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service